COM638467945 Combi-Blocks, Inc.

2-(Bis(2-chloroethyl)amino)ethanol hydrochloride

  • MDL number MFCD01677899
  • Cas Number : 63978-53-0
  • Molecular Weight : 222.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-0862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-0862
Availability details for
Quantity
  • Availability
  • Add to Cart