COM638468185 Combi-Blocks, Inc.

2-Methyloctan-1-ol

  • MDL number MFCD02258714
  • Cas Number : 818-81-5
  • Molecular Weight : 144.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-1834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-1834
Availability details for
Quantity
  • Availability
  • Add to Cart