COM638468298 Combi-Blocks, Inc.

3,4,5-Trihydroxybenzoic acid stearyl ester

  • MDL number MFCD00016429
  • Cas Number : 10361-12-3
  • Molecular Weight : 422.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-2303

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-2303
Availability details for
Quantity
  • Availability
  • Add to Cart