COM638468333 Combi-Blocks, Inc.

(S)-(-)-3-(Benzoylthio)-2-methylpropanoic acid

  • MDL number MFCD00038376
  • Cas Number : 72679-02-8
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-2437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-2437
Availability details for
Quantity
  • Availability
  • Add to Cart