COM638468609 Combi-Blocks, Inc.

3-Bromoquinoline-6-carbaldehyde

  • MDL number MFCD28665745
  • Cas Number : 860757-84-2
  • Molecular Weight : 236.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-3575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-3575
Availability details for
Quantity
  • Availability
  • Add to Cart