COM638468868 Combi-Blocks, Inc.

1,1'-(Oxybis(4,1-phenylene))bis(2-bromo-2-methylpropan-1-one)

  • MDL number MFCD29921811
  • Cas Number : 157891-77-5
  • Molecular Weight : 468.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-4634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-4634
Availability details for
Quantity
  • Availability
  • Add to Cart