COM638468922 Combi-Blocks, Inc.

5-Nitro-3-(piperazin-1-ylmethyl)-1h-indole

  • MDL number MFCD29920221
  • Cas Number : 1956324-74-5
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-4839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-4839
Availability details for
Quantity
  • Availability
  • Add to Cart