COM638469181 Combi-Blocks, Inc.

1-(Bromomethyl)-n-methylcyclopropanecarboxamide

  • MDL number MFCD29059143
  • Cas Number : 1956379-54-6
  • Molecular Weight : 192.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-5819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-5819
Availability details for
Quantity
  • Availability
  • Add to Cart