COM638469283 Combi-Blocks, Inc.

(2S,4S)-2-Amino-4-benzylpentanedioic acid hydrochloride

  • MDL number MFCD29920295
  • Cas Number : 169700-89-4
  • Molecular Weight : 273.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-6246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-6246
Availability details for
Quantity
  • Availability
  • Add to Cart