COM638469331 Combi-Blocks, Inc.

8-(Trifluoromethyl)chroman-4-amine

  • MDL number MFCD09026487
  • Cas Number : 890839-70-0
  • Molecular Weight : 217.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-6449

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-6449
Availability details for
Quantity
  • Availability
  • Add to Cart