COM638469524 Combi-Blocks, Inc.

(3-Methoxypyridazin-4-yl)methanol

  • MDL number MFCD29059152
  • Cas Number : 1591827-03-0
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-7239

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-7239
Availability details for
Quantity
  • Availability
  • Add to Cart