COM638469529 Combi-Blocks, Inc.

Methyl 3-((methylamino)methyl)benzoate

  • MDL number MFCD11858335
  • Cas Number : 922163-35-7
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-7252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-7252
Availability details for
Quantity
  • Availability
  • Add to Cart