COM638469721 Combi-Blocks, Inc.

4-Amino-2,6-dichloronicotinic acid

  • MDL number MFCD19105212
  • Cas Number : 929288-22-2
  • Molecular Weight : 207.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-7982

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-7982
Availability details for
Quantity
  • Availability
  • Add to Cart