COM638469740 Combi-Blocks, Inc.

(4-Methoxyphenyl)methanesulfonyl chloride

  • MDL number MFCD07778381
  • Cas Number : 110661-59-1
  • Molecular Weight : 220.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8067
Availability details for
Quantity
  • Availability
  • Add to Cart