COM638469793 Combi-Blocks, Inc.

2-(Adamantan-1-yl)-4-methyl-1h-pyrrole

  • MDL number MFCD29477676
  • Cas Number : 1914148-52-9
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8267

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8267
Availability details for
Quantity
  • Availability
  • Add to Cart