COM638469813 Combi-Blocks, Inc.

(1,4-Trans)-tert-butyl-4-(piperazin-1-yl)cyclohexyl)carbamate

  • MDL number MFCD29059870
  • Cas Number : 1956370-76-5
  • Molecular Weight : 283.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8388
Availability details for
Quantity
  • Availability
  • Add to Cart