COM638469828 Combi-Blocks, Inc.

1-((tert-Butyldimethylsilyl)oxy)-3-chloropropan-2-ol

  • MDL number MFCD29920232
  • Cas Number : 136917-95-8
  • Molecular Weight : 224.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8442
Availability details for
Quantity
  • Availability
  • Add to Cart