COM638469831 Combi-Blocks, Inc.

(1-Methyl-1h-benzo[d]imidazol-5-yl)methanol

  • MDL number MFCD11616105
  • Cas Number : 115576-91-5
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-8457
Availability details for
Quantity
  • Availability
  • Add to Cart