COM638470159 Combi-Blocks, Inc.

N'-((2S,3S)-2-(Benzyloxy)pentan-3-yl)formohydrazide oxalate

  • MDL number MFCD29044917
  • Cas Number : 1887197-42-3
  • Molecular Weight : 326.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-9856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-9856
Availability details for
Quantity
  • Availability
  • Add to Cart