COM638470230 Combi-Blocks, Inc.

Afatinib dimaleate

  • MDL number MFCD25974239
  • Cas Number : 850140-73-7
  • Molecular Weight : 718.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0235

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0235
Availability details for
Quantity
  • Availability
  • Add to Cart