COM638470235 Combi-Blocks, Inc.

3-Bromo-7-(methylsulfonyl)quinoline

  • MDL number MFCD29059318
  • Cas Number : 1956331-49-9
  • Molecular Weight : 286.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0314

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0314
Availability details for
Quantity
  • Availability
  • Add to Cart