COM638470287 Combi-Blocks, Inc.

3',5'-Di-o-acetyl-2'-deoxy-2'-fluoro-5-iodouridine

  • MDL number MFCD22124296
  • Cas Number : 565226-18-8
  • Molecular Weight : 456.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0618
Availability details for
Quantity
  • Availability
  • Add to Cart