COM638470393 Combi-Blocks, Inc.

4-(Chloromethyl)-5-methyl-2-(3,4,5-trimethoxyphenyl)-1,3-oxazole

  • MDL number MFCD10699135
  • Cas Number : 671216-07-2
  • Molecular Weight : 297.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1207
Availability details for
Quantity
  • Availability
  • Add to Cart