COM638470417 Combi-Blocks, Inc.

Ethyl 3-amino-4-phenylthiophene-2-carboxylate

  • MDL number MFCD12402771
  • Cas Number : 156424-61-2
  • Molecular Weight : 247.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1342

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1342
Availability details for
Quantity
  • Availability
  • Add to Cart