COM638470524 Combi-Blocks, Inc.

Methyl 3-(methyl([4-(propan-2-yl)phenyl]sulfonyl)amino)thiophene-2-carboxylate

  • MDL number MFCD19104881
  • Cas Number : 1325304-90-2
  • Molecular Weight : 353.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2100
Availability details for
Quantity
  • Availability
  • Add to Cart