COM638470535 Combi-Blocks, Inc.

6-Chloro-9-(2-o-methyl-beta-d-ribofuranosyl)-9h-purin-2-amine

  • MDL number MFCD28016446
  • Cas Number : 194034-59-8
  • Molecular Weight : 315.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2183
Availability details for
Quantity
  • Availability
  • Add to Cart