COM638470559 Combi-Blocks, Inc.

4-Bromobenzofuran-3-carbaldehyde

  • MDL number MFCD22035859
  • Cas Number : 1368302-72-0
  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2374
Availability details for
Quantity
  • Availability
  • Add to Cart