COM638470643 Combi-Blocks, Inc.

(2S)-Hydroxy(phenyl)acetic acid (2r)-n-benzyl-1-(4-methoxyphenyl)propan-2-amine

  • MDL number MFCD13194789
  • Cas Number : 188690-84-8
  • Molecular Weight : 407.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2914

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2914
Availability details for
Quantity
  • Availability
  • Add to Cart