COM638470702 Combi-Blocks, Inc.

[(5-Chloro-2-fluorophenyl)sulfonyl]acetonitrile

  • MDL number MFCD19705883
  • Cas Number : 1325305-64-3
  • Molecular Weight : 233.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3339

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3339
Availability details for
Quantity
  • Availability
  • Add to Cart