COM638470782 Combi-Blocks, Inc.

6-Amino-5-chloro-2-cyclopropylpyrimidine-4-carboxylic acid

  • MDL number MFCD17013133
  • Cas Number : 858956-08-8
  • Molecular Weight : 213.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3813
Availability details for
Quantity
  • Availability
  • Add to Cart