COM638470822 Combi-Blocks, Inc.

2-Phenylpropylene oxide

  • MDL number MFCD00059156
  • Cas Number : 2085-88-3
  • Molecular Weight : 134.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4096
Availability details for
Quantity
  • Availability
  • Add to Cart