COM638470848 Combi-Blocks, Inc.

Glyphosate isopropylamine salt

  • MDL number MFCD00075118
  • Cas Number : 38641-94-0
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4224
Availability details for
Quantity
  • Availability
  • Add to Cart