COM638470910 Combi-Blocks, Inc.

1,3-Dimethyl-2,4,5-trioxo-1,2,3,4,5,6,7,8-octahydropyrido[2,3-d]pyrimidine-7-carboxylic acid

  • MDL number MFCD18194602
  • Cas Number : 1260953-09-0
  • Molecular Weight : 253.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4659

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4659
Availability details for
Quantity
  • Availability
  • Add to Cart