COM638470918 Combi-Blocks, Inc.

4-Ethoxyquinoline-2-carboxylic acid

  • MDL number MFCD07636847
  • Cas Number : 40609-78-7
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4708

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4708
Availability details for
Quantity
  • Availability
  • Add to Cart