COM638471009 Combi-Blocks, Inc.

(2-Pyridylamino)methylenemalonic acid diethyl ester

  • MDL number MFCD00160377
  • Cas Number : 39080-52-9
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5289
Availability details for
Quantity
  • Availability
  • Add to Cart