COM638471012 Combi-Blocks, Inc.

2-Pyridin-4-ylpyrazolo[1,5-a]pyrazin-4(5h)-one

  • MDL number MFCD20528954
  • Cas Number : 1351398-75-8
  • Molecular Weight : 212.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5322

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5322
Availability details for
Quantity
  • Availability
  • Add to Cart