COM638471082 Combi-Blocks, Inc.

6,8-Difluoro-4-methoxyquinoline-2-carboxylic acid

  • MDL number MFCD21086556
  • Cas Number : 1338651-82-3
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5833
Availability details for
Quantity
  • Availability
  • Add to Cart