COM638471247 Combi-Blocks, Inc.

Pagoclone

  • MDL number MFCD00864700
  • Cas Number : 133737-32-3
  • Molecular Weight : 407.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6895
Availability details for
Quantity
  • Availability
  • Add to Cart