COM638471364 Combi-Blocks, Inc.

6-Fluoro-1-(4-methylphenyl)-1h-4,1,2-benzothiadiazine-3-carboxylic acid 4,4-dioxide

  • MDL number MFCD26130100
  • Cas Number : 1708401-67-5
  • Molecular Weight : 334.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7759
Availability details for
Quantity
  • Availability
  • Add to Cart