COM638471393 Combi-Blocks, Inc.

3'-Deoxy-3'-fluoro-5-methyluridine

  • MDL number MFCD27921028
  • Cas Number : 125217-37-0
  • Molecular Weight : 260.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7961

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7961
Availability details for
Quantity
  • Availability
  • Add to Cart