COM638471408 Combi-Blocks, Inc.

2-[(3-Fluoro-4-methoxyphenyl)amino]pyridine-3-sulfonamide

  • MDL number MFCD26130083
  • Cas Number : 1340753-74-3
  • Molecular Weight : 297.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8046
Availability details for
Quantity
  • Availability
  • Add to Cart