COM638471416 Combi-Blocks, Inc.

1-((2R,3S,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1h)-one

  • MDL number MFCD28976121
  • Cas Number : 136675-88-2
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8092
Availability details for
Quantity
  • Availability
  • Add to Cart