COM638471420 Combi-Blocks, Inc.

5-Oxo-1-phenyl-n-(1-[3-(3-thienyl)-1,2,4-oxadiazol-5-yl]cyclohexyl)pyrrolidine-3-carboxamide

  • MDL number MFCD22619640
  • Cas Number : 1396766-75-8
  • Molecular Weight : 436.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8117
Availability details for
Quantity
  • Availability
  • Add to Cart