COM638471427 Combi-Blocks, Inc.

5-(Methylsulfonyl)quinolin-3-amine

  • MDL number MFCD29059319
  • Cas Number : 1956341-17-5
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8165
Availability details for
Quantity
  • Availability
  • Add to Cart