COM638471434 Combi-Blocks, Inc.

Bis(4-(vinyloxy)butyl) succinate

  • MDL number MFCD01863595
  • Cas Number : 135876-32-3
  • Molecular Weight : 314.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8183
Availability details for
Quantity
  • Availability
  • Add to Cart