COM638471442 Combi-Blocks, Inc.

N-(5-Ethyl-1,3,4-thiadiazol-2-yl)acetamide

  • MDL number MFCD00500747
  • Cas Number : 33720-98-8
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8231

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8231
Availability details for
Quantity
  • Availability
  • Add to Cart