COM638471451 Combi-Blocks, Inc.

(2E,4E)-5-(4-Methoxy-2,3,6-trimethylphenyl)-3-methylpenta-2,4-dienal

  • MDL number MFCD12024630
  • Cas Number : 69877-38-9
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8293

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8293
Availability details for
Quantity
  • Availability
  • Add to Cart