COM638471452 Combi-Blocks, Inc.

1-(4-Aminobenzyl)-n-phenyl-1h-imidazole-4-carboxamide

  • MDL number MFCD20754680
  • Cas Number : 1351843-58-7
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8298
Availability details for
Quantity
  • Availability
  • Add to Cart