COM638471453 Combi-Blocks, Inc.

N6-Dimethylaminomethylidene isoguanosine

  • MDL number MFCD25542456
  • Cas Number : 156706-72-8
  • Molecular Weight : 338.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8308
Availability details for
Quantity
  • Availability
  • Add to Cart