COM638471454 Combi-Blocks, Inc.

(5-Methylpyridazin-4-yl)methanol

  • MDL number MFCD29059144
  • Cas Number : 1956324-99-4
  • Molecular Weight : 124.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8309
Availability details for
Quantity
  • Availability
  • Add to Cart